Sodium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Trifluoromethanesulphonic acid sodium salt

A148831-100G A148831-10G A148831-25G A148831-500G A148831-5G A148831-1000G
76719-660EA 78.51 USD
76719-660 76719-662 76719-664 76719-666 76719-668 77667-120
Sodium trifluoromethanesulfonate 98%
Sodium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: CF₃NaO₃S
MW: 172.06 g/mol
MDL Number: MFCD00061607
CAS Number: 2926-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR