2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 97%

Supplier: Ambeed
Warning

Synonyms: D-Fructopyranose diacetonide, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methanol, Diacetonefructose

A148534-100G A148534-25G A148534-10G A148534-5G
76719-642EA 44.56 USD
76719-642 76719-644 77567-080 77567-082
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 97%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose

Precaución: For research use only.

Formula: C₁₂H₂₀O₆
MW: 260.29 g/mol
MDL Number: MFCD00022183
CAS Number: 20880-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR