(4-(Phenylthio)phenyl)methanol 98%

Supplier: Ambeed
Warning

A148279-10G A148279-1G A148279-250MG A148279-500G A148279-5G
76719-616EA 12.74 USD
76719-616 76719-618 76719-620 76719-622 77667-104
(4-(Phenylthio)phenyl)methanol 98%
(4-(Phenylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₂OS
MW: 216.30 g/mol
MDL Number: MFCD04108413
CAS Number: 6317-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR