N2-Isopropyl-2,3-pyridinediamine 95%

Supplier: Ambeed
Warning

A147587-1G A147587-250MG
76719-592EA 124 USD
76719-592 76719-594
N2-Isopropyl-2,3-pyridinediamine 95%
N2-Isopropyl-2,3-pyridinediamine

Precaución: for Research Use Only

Formula: C₈H₁₃N₃
MDL Number: MFCD08692138
CAS Number: 24188-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR