(R)-Methyl 2-acetamido-3-mercaptopropanoate 97% (by HPLC)

Supplier: Ambeed
Danger

A147358-100MG A147358-25G A147358-1G A147358-250MG A147358-5G
76719-582EA 17 USD
76719-582 76719-584 77549-810 77600-098 77600-100
(R)-Methyl 2-acetamido-3-mercaptopropanoate 97% (by HPLC)
(R)-Methyl 2-acetamido-3-mercaptopropanoate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃S
MDL Number: MFCD00237476
CAS Number: 7652-46-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR