Biphenyl-4-sulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: Biphenyl-4-sulphonyl Chloride

A147186-10G A147186-25G A147186-25MG A147186-5G A147186-5MG
76719-564EA 11 USD
76719-564 76719-566 76719-568 76719-570 76719-572
Biphenyl-4-sulfonyl chloride 95%
Biphenyl-4-sulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₂H₉ClO₂S
MDL Number: MFCD00078658
CAS Number: 1623-93-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR