N-Isopropylacrylamide ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A147143-100G A147143-5G A147143-25G A147143-500G
76719-560EA 37.14 USD
76719-560 76719-562 77567-028 77592-754
N-Isopropylacrylamide ≥98% stabilized
N-Isopropylacrylamide

Precaución: For research use only.

Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00041913
CAS Number: 2210-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR