PP2 98%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butyl)-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

A146866-10MG A146866-1MG A146866-50MG A146866-5MG
76719-552EA 30.77 USD
76719-552 76719-554 76719-556 76719-558
PP2 98%
PP2

Precaución: for Research Use Only

Formula: C₁₅H₁₆ClN₅
MW: 301.8 g/mol
MDL Number: MFCD01568210
CAS Number: 172889-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR