Pyridoxine hydrochloride 95%

Supplier: Ambeed
Warning

A146452-100G A146452-100MG A146452-500G
76719-542EA 65 USD
76719-542 76719-544 76719-546 76719-548 76719-550
Pyridoxine hydrochloride 95%
Pyridoxine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO₃
MDL Number: MFCD00012807
CAS Number: 58-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR