Methyl thieno[3,2-b]thiophene-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A145881-100MG
A145881-1G
A145881-250MG
A145881-5G
76719-518EA
13
USD
76719-518
76719-520
76719-522
76719-524
Methyl thieno[3,2-b]thiophene-2-carboxylate 98%
Methyl thieno[3,2-b]thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₆O₂S₂ |
MDL Number:
MFCD01934498 CAS Number: 98800-10-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...