Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(1H-imidazol-1-yl)benzoate
A145728-10G
A145728-1G
A145728-250MG
A145728-25G
A145728-5G
76719-506EA
100.79
USD
76719-506
76719-508
76719-510
76719-512
76719-514
Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate 98%
Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O₂ MW: 202.21 g/mol |
MDL Number:
MFCD00127080 CAS Number: 101184-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...