Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%

Supplier: Ambeed
Warning

Synonyms: Sodium phytate, Dodecasodium (2,3,4,5,6-pentaphosphonatooxycyclohexyl) phosphate

A145326-1000G A145326-100G A145326-10G A145326-500G
76719-488EA 155.96 USD
76719-488 76719-490 76719-492 76719-494
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate)

Precaución: for Research Use Only

Formula: C₆H₆Na₁₂O₂₄P₆
MW: 923.81 g/mol
MDL Number: MFCD00082310
CAS Number: 14306-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR