N,N-Dimethylpiperazine-1-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Dimethylcarbamoyl)piperazine

A144876-1G A144876-250MG A144876-5G
76719-474EA 99.73 USD
76719-474 76719-476 77592-714
N,N-Dimethylpiperazine-1-carboxamide 97%
N,N-Dimethylpiperazine-1-carboxamide

Precaución: for Research Use Only

Formula: C₇H₁₅N₃O
MW: 157.213 g/mol
MDL Number: MFCD01050443
CAS Number: 41340-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR