Fmoc-Thi-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-3-(2-thienyl)-L-alanine, Fmoc-β-(2-thienyl)-Ala-OH, (S)-2-(Fmoc-amino)-3-(2-thienyl)propionic acid, N-α-Fmoc-β-thienyl-L-alanine
A144819-10G
A144819-1G
A144819-250MG
A144819-25G
A144819-5G
76719-464EA
194.15
USD
76719-464
76719-466
76719-468
76719-470
76719-472
Fmoc-Thi-OH 95%
Fmoc-Thi-OH
Precaución: for Research Use Only
Formula:
C₂₂H₁₉NO₄S MW: 393.47 g/mol |
MDL Number:
MFCD00065676 CAS Number: 130309-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...