Diphenylphosphinic acid 98%

Supplier: Ambeed
Warning

A144127-100G A144127-10G A144127-25G A144127-500G A144127-5G
76719-434EA 36.08 USD
76719-434 76719-436 76719-438 76719-440 76719-442
Diphenylphosphinic acid 98%
Diphenylphosphinic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁O₂P
MW: 218.19 g/mol
MDL Number: MFCD00002132
CAS Number: 1707-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR