4-(Diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate 98%

Supplier: Ambeed
Warning

A144038-100MG A144038-10G A144038-1MG A144038-250MG A144038-50MG
76719-424EA 21.22 USD
76719-424 76719-426 76719-428 76719-430 76719-432
4-(Diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate 98%
4-(Diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate

Precaución: for Research Use Only

Formula: C₂₂H₃₁NO₃
MW: 357.48 g/mol
MDL Number: MFCD00865252
CAS Number: 5633-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR