4-(Piperidin-1-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A143957-1G A143957-250MG A143957-25G
76719-422EA 10 USD
76719-422 77879-013 77879-014
4-(Piperidin-1-yl)benzaldehyde 98%
4-(Piperidin-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MDL Number: MFCD00778395
CAS Number: 10338-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR