4-(Trifluoromethyl)phenylboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4,5,5-Tetramethyl-2-(4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 4-Trifluoromethylphenylboronic acid pinacol ester
A143941-100G
A143941-25G
A143941-5G
A143941-1G
76719-416EA
392.54
USD
76719-416
76719-418
76719-420
77600-076
4-(Trifluoromethyl)phenylboronic acid pinacol ester 97%
4-(Trifluoromethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BF₃O₂ MW: 272.0712 g/mol |
MDL Number:
MFCD05863924 CAS Number: 214360-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...