Methyl trans-p-coumarate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl trans-4-Hydroxycinnamate, Methyl (E)-3-(4-hydroxyphenyl)acrylate, trans-4-Hydroxycinnamic Acid Methyl Ester, trans-p-Coumaric Acid Methyl Ester, (E)-Methyl 4-coumarate

A141299-100G A141299-100MG A141299-25MG A141299-5G A141299-25G
76719-370EA 151.71 USD
76719-370 76719-372 76719-374 76719-376 77600-052
Methyl trans-p-coumarate 98%
Methyl trans-p-coumarate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD00157167
CAS Number: 19367-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR