4-Cyclohexylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenylcyclohexane

A141120-10G A141120-25G
76719-364EA 14.86 USD
76719-364 76719-366
4-Cyclohexylphenol 98%
4-Cyclohexylphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD00035699
CAS Number: 1131-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR