N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

A140258-5G A140258-1G A140258-250MG A140258-25G
76719-332EA 380 USD
76719-332 77600-040 77600-042 77878-989
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 97%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
MDL Number: MFCD00063682
CAS Number: 21653-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR