N-(3-Hydroxypropyl)phthalimide 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Hydroxypropyl)isoindoline-1,3-dione

A139927-10G A139927-1G A139927-5G A139927-100G A139927-25G
76719-316EA 14 USD
76719-316 76719-318 76719-320 77592-658 77592-660
N-(3-Hydroxypropyl)phthalimide 95%
N-(3-Hydroxypropyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00023097
CAS Number: 883-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR