6-Ethoxy-2-naphthaleneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Ethoxynaphthalen-2-yl)boronic acid

A139473-10G A139473-250MG A139473-5G
76719-300EA 81.69 USD
76719-300 76719-302 76719-304
6-Ethoxy-2-naphthaleneboronic acid 98%
6-Ethoxy-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃BO₃
MW: 216.04 g/mol
MDL Number: MFCD03427009
CAS Number: 352525-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR