4-Pyrid-3-ylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridin-3-yl-benzoic acid

A139418-100MG A139418-1G A139418-250MG A139418-25G A139418-5G
76719-290EA 7.43 USD
76719-290 76719-292 76719-294 76719-296 76719-298
4-Pyrid-3-ylbenzoic acid 97%
4-Pyrid-3-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD03426513
CAS Number: 4385-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR