Potassium (3-(methanesulfonylamino)phenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(3-(methylsulfonamido)phenyl)borate

A138949-1G A138949-250MG A138949-5G
76719-282EA 55 USD
76719-282 76719-284 76719-286
Potassium (3-(methanesulfonylamino)phenyl)trifluoroborate 97%
Potassium (3-(methanesulphonylamino)phenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₈BF₃KNO₂S
MW: 277.11 g/mol
MDL Number: MFCD04115762
CAS Number: 850623-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR