Methyl-3,5-bis(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,5-bis(trifluoromethyl)benzoate

A138809-10G A138809-1G A138809-100G
76719-278EA 18.04 USD
76719-278 76719-280 77592-636
Methyl-3,5-bis(trifluoromethyl)benzoate 98%
Methyl 3,5-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₆F₆O₂
MW: 272.15 g/mol
MDL Number: MFCD00052317
CAS Number: 26107-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR