3-(4-Nitrophenyl)-2-oxopropanoic acid 97%

Supplier: Ambeed
Warning

A138262-1G A138262-250MG A138262-5G A138262-100MG
76719-272EA 101 USD
76719-272 76719-274 76719-276 77878-976
3-(4-Nitrophenyl)-2-oxopropanoic acid 97%
3-(4-Nitrophenyl)-2-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₅
MDL Number: MFCD00031314
CAS Number: 38335-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR