Methyl-3-hydroxy-4-iodobenzoate 97%
Supplier: Ambeed
|
Warning
|
A138032-100MG
A138032-10G
A138032-1G
A138032-250MG
A138032-25G
A138032-5G
76719-260EA
5
USD
76719-260
76719-262
76719-264
76719-266
76719-268
76719-270
Methyl-3-hydroxy-4-iodobenzoate 97%
Methyl-3-hydroxy-4-iodobenzoate
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ |
MDL Number:
MFCD06203960 CAS Number: 157942-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...