Mubritinib ≥98%

Supplier: Ambeed
Warning

Synonyms: (E)-4-[[4-[4-(1H-1,2,3-Triazol-1-yl)butyl]phenoxy]methyl]-2-[4-(trifluoromethyl)styryl]oxazole, TAK 165, (E)-4-((4-(4-(1H-1,2,3-Triazol-1-yl)butyl)phenoxy)methyl)-2-(4-(trifluoromethyl)styryl)oxazole

A137400-1MG A137400-10MG A137400-5MG
76719-250EA 20.16 USD
76719-250 77666-984 77666-986
Mubritinib ≥98%
Mubritinib

Precaución: for Research Use Only

Formula: C₂₅H₂₃F₃N₄O₂
MW: 468.5 g/mol
MDL Number: MFCD09954135
CAS Number: 366017-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR