2,8-Dibromdibenzothiophene 97%

Supplier: Ambeed
Warning

A137111-100MG A137111-10G A137111-1G A137111-250MG A137111-25G A137111-5G A137111-100G
76719-236EA 5 USD
76719-236 76719-238 76719-240 76719-242 76719-244 76719-246 77878-972
2,8-Dibromdibenzothiophene 97%
2,8-Dibromdibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂S
MDL Number: MFCD00092755
CAS Number: 31574-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR