N,N'-(2-Chloro-1,4-phenylene)diacetamide 98%

Supplier: Ambeed
Warning

A137099-100MG A137099-250MG A137099-5G
76719-230EA 17 USD
76719-230 76719-232 76719-234
N,N'-(2-Chloro-1,4-phenylene)diacetamide 98%
N,N'-(2-Chloro-1,4-phenylene)diacetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
MDL Number: MFCD00221890
CAS Number: 50610-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR