S-Adenosyl-L-methionine disulfate tosylate 95%

Supplier: Ambeed
Warning

Synonyms: ((S)-3-Amino-3-carboxypropyl)(((2S,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)(methyl)sulfonium hydrogen sulfate 4-methylbenzenesulfonate sulfate

A136497-100MG A136497-10G A136497-100G A136497-500G
76719-212EA 5 USD
76719-212 77592-588 77599-984 77599-986
S-Adenosyl-L-methionine disulfate tosylate 95%
S-Adenosyl-L-methionine disulfate tosylate

Precaución: for Research Use Only

Formula: C₂₂H₃₄N₆O₁₆S₄
MW: 766.8 g/mol
MDL Number: MFCD01674068
CAS Number: 97540-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR