Pitavastatin calcium 98%

Supplier: Ambeed
Warning

A135712-100MG A135712-10MG A135712-250MG A135712-25G A135712-50MG A135712-5G A135712-1G A135712-1MG
76719-162EA 22 USD
76719-162 76719-164 76719-166 76719-168 76719-170 76719-172 77566-674 77666-958
Pitavastatin calcium 98%
Pitavastatin calcium

Precaución: For research use only.

Formula: C₅₀H₄₆CaF₂N₂O₈
MDL Number: MFCD01937979
CAS Number: 147526-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR