Potassium(4-nitrophenyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(4-nitrophenyl)borate, (4-Nitrophenyl)trifluoroboric acid potassium salt

A135383-1G A135383-250MG A135383-5G
76719-146EA 14 USD
76719-146 76719-148 76719-150
Potassium(4-nitrophenyl)trifluoroborate 95%
Potassium(4-nitrophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BF₃KNO₂
MW: 229.01
MDL Number: MFCD04115766
CAS Number: 850623-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR