5-Bromo-2-nitrophenol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-3-hydroxy-4-nitrobenzene

A135322-100G A135322-10G A135322-1G A135322-25G A135322-5G
76719-136EA 195 USD
76719-136 76719-138 76719-140 76719-142 76719-144
5-Bromo-2-nitrophenol 98%
5-Bromo-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MDL Number: MFCD03095027
CAS Number: 27684-84-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR