Indole-3-butyric acid potassium salt 97%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-(1H-indol-3-yl)butanoate

A134975-1000G A134975-100G A134975-10G A134975-25G A134975-500G A134975-5G
76719-102EA 408.45 USD
76719-102 76719-104 76719-106 76719-108 76719-110 76719-112
Indole-3-butyric acid potassium salt 97%
Indole-3-butyric acid potassium salt

Precaución: for Research Use Only

Formula: C₁₂H₁₂KNO₂
MW: 241.331 g/mol
MDL Number: MFCD00058442
CAS Number: 60096-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR