N4-Acetylsulfamethoxazole 98+%

Supplier: Ambeed
Warning

Synonyms: N-(4-(N-(5-Methylisoxazol-3-yl)sulphamoyl)phenyl)acetamide

A134633-100MG A134633-1G A134633-250MG A134633-5G
76719-086EA 25.47 USD
76719-086 76719-088 76719-090 76719-092
N4-Acetylsulfamethoxazole 98+%
N4-Acetylsulfamethoxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₄S
MW: 295.3143 g/mol
MDL Number: MFCD00205417
CAS Number: 21312-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR