n-Octylamine hydrobromide 98%

Supplier: Ambeed
Warning

A134370-1G A134370-25G A134370-5G
76719-080EA 21 USD
76719-080 76719-082 76719-084
n-Octylamine hydrobromide 98%
n-Octylamine hydrobromide

Precaución: for Research Use Only

Formula: C₈H₁₉N·HBr
MDL Number: MFCD00044802
CAS Number: 14846-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR