Nifedipine 98%

Supplier: Ambeed
Warning

A134096-100G A134096-1G A134096-25G A134096-500G A134096-5G A134096-1000G
76719-050EA 100 USD
76719-050 76719-052 76719-054 76719-056 76719-058 77666-932
Nifedipine 98%
Nifedipine

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₆
MDL Number: MFCD00057326
CAS Number: 21829-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR