N-Methyl-D-alanine 97%

Supplier: Ambeed
Warning

Synonyms: N-D-Me-Ala-OH, H-N-Me-D-Ala-OH

A133411-1G A133411-250MG A133411-10G A133411-5G A133411-100G A133411-25G
76719-032EA 11.67 USD
76719-032 76719-034 77599-916 77599-918 77666-918 77666-920
N-Methyl-D-alanine 97%
N-Methyl-D-alanine

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.121 g/mol
MDL Number: MFCD00063135
CAS Number: 29475-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR