Quinacridone, Technical Grade

Supplier: Ambeed
Warning

Synonyms: 5,12-Dihydroquino[2,3-b]acridine-7,14-dione, Pigment Violet 19

A132462-100G A132462-5G A132462-25G A132462-500G
76719-008EA 103 USD
76719-008 76719-010 77592-522 77666-902
Quinacridone, Technical Grade
Quinacridone

Precaución: for Research Use Only

Formula: C₂₀H₁₂N₂O₂
MDL Number: MFCD00059956
CAS Number: 1047-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR