Methyl phenylpropiolate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl Phenylpropiolate, Phenylpropiolic acid methyl ester

A132234-100G A132234-10G A132234-1G A132234-25G A132234-5G
76718-996EA 450.89 USD
76718-996 76718-998 76719-000 76719-002 76719-004
Methyl phenylpropiolate 98%
Methyl phenylpropiolate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00041685
CAS Number: 4891-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR