4-Chloropyrido[3,2-d]pyrimidine 98%

Supplier: Ambeed
Warning

A132138-1G A132138-250MG A132138-5G
76718-990EA 143 USD
76718-990 76718-992 76718-994
4-Chloropyrido[3,2-d]pyrimidine 98%
4-Chloropyrido[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄ClN₃
MW: 165.5798 g/mol
MDL Number: MFCD10566748
CAS Number: 51674-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR