N-Hydroxy-1-hydrazinecarboxamide 98+%

Supplier: Ambeed
Danger

A131431-1G A131431-5G
76718-978EA 26 USD
76718-978 77878-947
N-Hydroxy-1-hydrazinecarboxamide 98+%
N-Hydroxy-1-hydrazinecarboxamide

Precaución: for Research Use Only

Formula: CH₅N₃O₂
MDL Number: MFCD03791199
CAS Number: 21520-79-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR