1-Phenylicosane-1,3-dione 95%

Supplier: Ambeed
Warning

A131384-1G A131384-250MG A131384-5G
76718-974EA 44 USD
76718-974 76718-976 77666-888
1-Phenylicosane-1,3-dione 95%
1-Phenylicosane-1,3-dione

Precaución: for Research Use Only

Formula: C₂₆H₄₂O₂
MDL Number: MFCD08274657
CAS Number: 58446-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR