N,N,N'-Trimethyl-1,3-propanediamine 98%

Supplier: Ambeed
Danger

Synonyms: N,n,n-trimethyl-1,3-propanediamine

A131332-1G A131332-25G A131332-5G
76718-956EA 10.61 USD
76718-956 76718-958 76718-960
N,N,N'-Trimethyl-1,3-propanediamine 98%
N,N,N'-Trimethyl-1,3-propanediamine

Precaución: for Research Use Only

Formula: C₆H₁₆N₂
MW: 116.208 g/mol
MDL Number: MFCD00014879
CAS Number: 4543-96-8
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR