4-(4-Hydroxypiperidin-1-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A131256-1G A131256-250MG A131256-5G
76718-944EA 41 USD
76718-944 76718-946 76718-948
4-(4-Hydroxypiperidin-1-yl)benzaldehyde 95%
4-(4-Hydroxypiperidin-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD05864666
CAS Number: 79421-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR