2-Nonynoic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl non-2-ynoate, Methyl 2-nonynoate

A130972-100G A130972-25G A130972-500G A130972-5G
76718-936EA 100.79 USD
76718-936 76718-938 76718-940 76718-942
2-Nonynoic acid methyl ester 95%
2-Nonynoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
MDL Number: MFCD00009547
CAS Number: 111-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR