2-(1-((Benzyloxy)carbonyl)piperidin-4-yl)acetic acid 95%

Supplier: Ambeed
Warning

A130529-100MG A130529-10G A130529-1G A130529-250MG A130529-5G
76718-924EA 5.31 USD
76718-924 76718-926 76718-928 76718-930 76718-932
2-(1-((Benzyloxy)carbonyl)piperidin-4-yl)acetic acid 95%
2-(1-((Benzyloxy)carbonyl)piperidin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MW: 277.3200 g/mol
MDL Number: MFCD02179001
CAS Number: 63845-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR