Methyl-5-chloro-2-cyanobenzoate 97%

Supplier: Ambeed
Warning

A130502-1G A130502-250MG A130502-5G
76718-920EA 27 USD
76718-920 76718-922 77878-940
Methyl-5-chloro-2-cyanobenzoate 97%
Methyl-5-chloro-2-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD08234733
CAS Number: 439117-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR